The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000274.3 | NP_000265.1 | 7 - 8 | 1034 | 98 |
NM_001171814.1 | NP_001165285.1 | 6 - 7 | 806 | 98 | |
NM_001322965.1 | NP_001309894.1 | 7 - 8 | 1029 | 98 | |
NM_001322966.1 | NP_001309895.1 | 8 - 9 | 1349 | 98 | |
NM_001322967.1 | NP_001309896.1 | 8 - 9 | 1154 | 98 | |
NM_001322968.1 | NP_001309897.1 | 9 - 10 | 1241 | 98 | |
NM_001322969.1 | NP_001309898.1 | 8 - 9 | 1121 | 98 | |
NM_001322970.1 | NP_001309899.1 | 9 - 10 | 1337 | 98 | |
NM_001322971.1 | NP_001309900.1 | 5 - 6 | 713 | 98 | |
NM_001322974.1 | NP_001309903.1 | 7 - 8 | 920 | 98 | |
XM_017016279.1 | XP_016871768.1 | 7 - 8 | 2829 | 98 | |
GenBank mRNA | AB208817.1 | - | 7 - 8 | 2819 | 98 |
AK296032.1 | - | 6 - 7 | 753 | 98 | |
AK312561.1 | - | 7 - 8 | 981 | 98 | |
AK315947.1 | - | 6 - 7 | 753 | 98 | |
BC000964.2 | - | 7 - 8 | 962 | 98 | |
BC016928.1 | - | 7 - 8 | 966 | 98 | |
CR457045.1 | - | 6 - 7 | 901 | 98 | |
CR749808.1 | - | 6 - 7 | 756 | 98 | |
M12267.1 | - | 7 - 8 | 955 | 98 | |
M14963.1 | - | 6 - 7 | 945 | 98 | |
M23204.1 | - | 7 - 8 | 952 | 98 | |
Y07511.1 | - | 6 - 7 | 941 | 98 |