Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXD4L6
Entrez Gene ID
653404
Gene Name
forkhead box D4-like 6
Gene Aliases
-
Chromosome Location
Chr.9: 41126251 - 41128975 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085476.1 NP_001078945.1 1 - 1 1656 141
GenBank mRNA BC103887.2 - 1 - 1 1064 141
BC137365.1 - 1 - 1 1144 141
BC137366.1 - 1 - 1 1144 141

Gene Symbol
FOXD4L3
Entrez Gene ID
286380
Gene Name
forkhead box D4-like 3
Gene Aliases
FOXD4L2, FOXD6
Chromosome Location
Chr.9: 68302867 - 68305084 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199135.4 NP_954586.4 1 - 1 1149 141
GenBank mRNA AY344641.1 - 1 - 1 1149 141
AY344642.1 - 1 - 1 1149 141

Gene Symbol
FOXD4L5
Entrez Gene ID
653427
Gene Name
forkhead box D4-like 5
Gene Aliases
bA15J10.2
Chromosome Location
Chr.9: 65282101 - 65285209 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126334.1 NP_001119806.1 1 - 1 1896 141

View Full Product Details