Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNRPN
Entrez Gene ID
6638
Gene Name
small nuclear ribonucleoprotein polypeptide N
Gene Aliases
HCERN3, PWCR, RT-LI, SM-D, SMN, SNRNP-N, SNURF-SNRPN, sm-N
Chromosome Location
Chr.15: 24823637 - 24978723 on Build GRCh38
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF400499.1 - LIT2210
IC-SNURF-SNRPN 3 prime noncoding transcripts
5 - 6 407 60
AF400501.1 - LIT2214
IC-SNURF-SNRPN 3 prime noncoding transcripts
1 - 2 626 60


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small nucleolar RNA, C/D box 115-27 SNORD115-27 HBII-52-27 Sense Chr. - 25220503 - 25220578 on Build GRCh38
Downstream small nucleolar RNA, C/D box 115-28 SNORD115-28 HBII-52-28 64.0 Sense Chr. - 25222354 - 25222427 on Build GRCh38

View Full Product Details