The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
GenBank mRNA | AY927425.1 | - | LIT3464 AY927425 |
1 - 1 | 253 | 134 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | GTSE1 antisense RNA 1 (head to head) | GTSE1-AS1 | CN5H6.4, LL22NC03-5H6.6 | -32.4kb | Anti-sense | Chr. - 46295143 - 46296660 on Build GRCh38 |
Downstream | tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase | TRMU | LCAL3, MTO2, MTU1, TRMT, TRMT1, TRNT1 | 5.8kb | Sense | Chr. - 46335401 - 46357340 on Build GRCh38 |