Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMX2OS
Entrez Gene ID
196047
Gene Name
EMX2 opposite strand/antisense RNA
Gene Aliases
EMX2-AS1, NCRNA00045
Chromosome Location
Chr.10: 117484293 - 117545068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002791.2 2 - 3 580 118
GenBank mRNA AY117034.1 - 2 - 3 580 118


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream PDZ domain containing 8 PDZD8 PDZK8 -116.6kb Sense Chr. - 117280489 - 117375467 on Build GRCh38
Downstream RAB11 family interacting protein 2 RAB11FIP2 Rab11-FIP2, nRip11 460.5kb Sense Chr. - 118004916 - 118046921 on Build GRCh38

View Full Product Details