Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HYMAI
- Entrez Gene ID
-
57061
- Gene Name
- hydatidiform mole associated and imprinted (non-protein coding)
- Gene Aliases
-
NCRNA00020
- Chromosome Location
-
Chr.6: 144004916 - 144008730 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF241534.1 |
- |
1 - 1 |
4304 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW300689.1 |
- |
1 |
|
|
Chr. 6 - 144004199 - 144004602 [+] on Build GRCh38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger C2HC-type containing 1B |
ZC2HC1B |
C6orf94, FAM164B, dJ468K18.5 |
-65.9kb
|
Sense |
Chr. - 143864436 - 143938346 on Build GRCh38 |
Downstream |
splicing factor 3b subunit 5 |
SF3B5 |
SF3b10, Ysf3 |
90.3kb
|
Anti-sense |
Chr. - 144094881 - 144095617 on Build GRCh38 |
View Full Product Details