Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP20-4
Entrez Gene ID
100151643
Gene Name
keratin associated protein 20-4
Gene Aliases
KAP20.4
Chromosome Location
Chr.21: 30620627 - 30620850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_023342.1 1 - 1 144 80

Gene Symbol
SPATA31C2
Entrez Gene ID
645961
Gene Name
SPATA31 subfamily C member 2
Gene Aliases
FAM75C2
Chromosome Location
Chr.9: 88129305 - 88134985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB096956.1 - 1 - 1 144 80


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream keratin associated protein 20-1 KRTAP20-1 KAP20.1 -4.0kb Sense Chr. - 30616455 - 30616625 on Build GRCh38
Downstream keratin associated protein 20-2 KRTAP20-2 KAP20.2 14.4kb Sense Chr. - 30635264 - 30635461 on Build GRCh38

View Full Product Details