Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTX2P1-UPK3BP1-PMS2P11
Entrez Gene ID
441263
Gene Name
DTX2P1-UPK3BP1-PMS2P11 readthrough, transcribed pseudogene
Gene Aliases
PMS2L11
Chromosome Location
Chr.7: 76980822 - 77023761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_023383.1 8 - 9 1322 87
GenBank mRNA BG982651.1 - 1 - 2 87

Gene Symbol
PMS2P10
Entrez Gene ID
100288713
Gene Name
PMS1 homolog 2, mismatch repair system component pseudogene 10
Gene Aliases
PMS2L10, PMSR4
Chromosome Location
Chr.7: 75324481 - 75327776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC047753.1 - 3 - 4 257 87

Gene Symbol
PMS2P2
Entrez Gene ID
5380
Gene Name
PMS1 homolog 2, mismatch repair system component pseudogene 2
Gene Aliases
PMS2L2, PMS4
Chromosome Location
Chr.7: 75343937 - 75358997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003614.3 3 - 4 281 87
GenBank mRNA DA215155.1 - 3 - 4 268 87

Gene Symbol
PMS2P5
Entrez Gene ID
5383
Gene Name
PMS1 homolog 2, mismatch repair system component pseudogene 5
Gene Aliases
PMS2L5, PMS7
Chromosome Location
Chr.7: 74890768 - 74921138 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BF984173.1 - 3 - 4 305 87
D38439.1 - 2 - 3 218 87
D38501.1 - 2 - 3 218 87

Gene Symbol
PMS2P9
Entrez Gene ID
100132832
Gene Name
PMS1 homolog 2, mismatch repair system component pseudogene 9
Gene Aliases
PMS2L17, PMS2LP1, PMSR5
Chromosome Location
Chr.7: 77039480 - 77053038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028058.1 2 - 3 697 87

View Full Product Details