Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WT1-AS
Entrez Gene ID
51352
Gene Name
WT1 antisense RNA
Gene Aliases
WIT-1, WIT1, WT1-AS1, WT1AS
Chromosome Location
Chr.11: 32435518 - 32458769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_023920.2 1 - 2 330 120
GenBank mRNA DB262871.1 - 1 - 2 109 120


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX394943.2 - 1 - 2 34 Chr. 11 - 32435814 - 32438749 [+] on Build GRCh38 120
GenBankmRNA DA952652.1 - 1 - 2 286 Chr. 11 - 32435565 - 32438063 [+] on Build GRCh38 120
GenBankmRNA DA961425.1 - 1 - 2 106 Chr. 11 - 32435744 - 32438316 [+] on Build GRCh38 120
GenBankmRNA BI837715.1 - 1 - 2 271 Chr. 11 - 32435579 - 32438389 [+] on Build GRCh38 123
GenBankmRNA DB192632.1 - 1 - 2 61 Chr. 11 - 32435787 - 32438354 [+] on Build GRCh38 120
GenBankmRNA DB269002.1 - 1 - 2 38 Chr. 11 - 32435810 - 32438370 [+] on Build GRCh38 120
GenBankmRNA BM545678.1 - 194 Chr. - - [-] on Build 120
GenBankmRNA BM906117.1 - 204 Chr. - - [-] on Build 120
GenBankmRNA DA704288.1 - 1 - 2 38 Chr. 11 - 32435810 - 32438303 [+] on Build GRCh38 120


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small nucleolar RNA, H/ACA box 88 SNORA88 -291.6kb Anti-sense Chr. - 32143742 - 32143912 on Build GRCh38
Downstream protein GVQW2-like LOC107984351 37.1kb Anti-sense Chr. - 32475731 - 32483540 on Build GRCh38

View Full Product Details