The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024113.1 | 1 - 2 | 319 | 69 | |
GenBank mRNA | AF387612.1 | - | 1 - 2 | 280 | 69 |
AK126929.1 | - | 1 - 2 | 319 | 69 | |
AY486332.1 | - | 1 - 2 | 289 | 69 | |
DA214065.1 | - | 1 - 2 | 319 | 69 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | synapse defective Rho GTPase homolog 2 | SYDE2 | -57.2kb | Sense | Chr. - 85157673 - 85201725 on Build GRCh38 | |
Downstream | B-cell CLL/lymphoma 10 | BCL10 | CARMEN, CIPER, CLAP, IMD37, c-E10, mE10 | 6.1kb | Sense | Chr. - 85265776 - 85276904 on Build GRCh38 |