The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_126169.1 | 2 - 3 | 1735 | 95 | |
GenBank mRNA | AK094767.1 | - | 2 - 3 | 1735 | 95 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | sodium voltage-gated channel alpha subunit 4 | SCN4A | CMS16, HOKPP2, HYKPP, HYPP, NAC1A, Na(V)1.4, Nav1.4, SkM1 | -5.9kb | Sense | Chr. - 63938554 - 63972918 on Build GRCh38 |
Downstream | proline rich 29 | PRR29 | C17orf72 | 422.0 | Anti-sense | Chr. - 63998351 - 64004305 on Build GRCh38 |