Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATP5J2
- Entrez Gene ID
-
9551
- Gene Name
- ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2
- Gene Aliases
-
ATP5JL
- Chromosome Location
-
Chr.7: 99458161 - 99466201 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK123419.1 |
- |
1 - 1 |
2747 |
80 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU680821.1 |
- |
1 |
|
|
Chr. 7 - 99458195 - 99458905 [+] on Build GRCh38 |
80 |
GenBankmRNA |
BM980489.1 |
- |
1 |
|
|
Chr. 7 - 99458195 - 99458942 [+] on Build GRCh38 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ATP5J2-PTCD1 readthrough |
ATP5J2-PTCD1 |
|
|
Sense |
Chr. - 99416739 - 99466201 on Build GRCh38 |
Downstream |
zinc finger protein 789 |
ZNF789 |
|
11.3kb
|
Anti-sense |
Chr. - 99472819 - 99487600 on Build GRCh38 |
View Full Product Details