Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DCAF12
- Entrez Gene ID
-
25853
- Gene Name
- DDB1 and CUL4 associated factor 12
- Gene Aliases
-
CT102, KIAA1892, TCC52, WDR40A
- Chromosome Location
-
Chr.9: 34086383 - 34126773 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF085907.1 |
- |
1 - 1 |
252 |
90 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
R98118.1 |
- |
|
246 |
|
Chr. - - [-] on Build |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ubiquitin associated protein 2 |
UBAP2 |
UBAP-2 |
-58.2kb
|
Anti-sense |
Chr. - 33921693 - 34048949 on Build GRCh38 |
Downstream |
ubiquitin associated protein 1 |
UBAP1 |
NAG20, UAP, UBAP, UBAP-1 |
71.3kb
|
Sense |
Chr. - 34179005 - 34252523 on Build GRCh38 |
View Full Product Details