Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CLLU1
- Entrez Gene ID
-
574028
- Gene Name
- chronic lymphocytic leukemia up-regulated 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 92421531 - 92431002 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ845167.1 |
- |
1 - 1 |
1845 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101928617 |
LOC101928617 |
|
-254.0kb
|
Sense |
Chr. - 92146085 - 92185784 on Build GRCh38 |
Downstream |
uncharacterized LOC100507616 |
LOC100507616 |
|
42.5kb
|
Sense |
Chr. - 92466440 - 92488106 on Build GRCh38 |
View Full Product Details