Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PCA3
- Entrez Gene ID
-
50652
- Gene Name
- prostate cancer associated 3 (non-protein coding)
- Gene Aliases
-
DD3, NCRNA00019, PCAT3
- Chromosome Location
-
Chr.9: 76764436 - 76787569 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
DQ374660.1 |
- |
1 - 2 |
47 |
101 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
prune homolog 2 |
PRUNE2 |
BMCC1, BNIPXL, C9orf65, KIAA0367 |
|
Anti-sense |
Chr. - 76611376 - 76906220 on Build GRCh38 |
Downstream |
forkhead box B2 |
FOXB2 |
bA159H20.4 |
235.5kb
|
Sense |
Chr. - 77019655 - 77020953 on Build GRCh38 |
View Full Product Details