Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FCF1
- Entrez Gene ID
-
51077
- Gene Name
- FCF1 rRNA-processing protein
- Gene Aliases
-
Bka, C14orf111, CGI-35, UTP24
- Chromosome Location
-
Chr.14: 74713118 - 74738620 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA032623.1 |
- |
1 |
420 |
|
Chr. 14 - 74736490 - 74737067 [+] on Build GRCh38 |
64 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Downstream |
YLP motif containing 1 |
YLPM1 |
C14orf170, PPP1R169, ZAP113, ZAP3 |
24.7kb
|
Sense |
Chr. - 74763322 - 74837310 on Build GRCh38 |
View Full Product Details