Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PIGT
- Entrez Gene ID
-
51604
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class T
- Gene Aliases
-
CGI-06, MCAHS3, NDAP, PNH2
- Chromosome Location
-
Chr.20: 45416067 - 45426245 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF305815.2 |
- |
1 - 1 |
1718 |
94 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI061662.1 |
- |
1 |
|
|
Chr. 20 - 45421354 - 45422032 [+] on Build GRCh38 |
94 |
GenBankmRNA |
AW370560.1 |
- |
1 |
|
|
Chr. 20 - 45421434 - 45422154 [+] on Build GRCh38 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SYS1-DBNDD2 readthrough (NMD candidate) |
SYS1-DBNDD2 |
C20orf169-DBNDD2 |
-10.7kb
|
Anti-sense |
Chr. - 45363169 - 45410610 on Build GRCh38 |
Downstream |
microRNA 6812 |
MIR6812 |
hsa-mir-6812 |
2.0kb
|
Anti-sense |
Chr. - 45425510 - 45425573 on Build GRCh38 |
View Full Product Details