Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGCC
Entrez Gene ID
28984
Gene Name
regulator of cell cycle
Gene Aliases
C13orf15, RGC-32, RGC32, bA157L14.2
Chromosome Location
Chr.13: 41457406 - 41470877 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK091099.1 - 1 - 1 2048 80


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA171994.1 - 1 141 Chr. 13 - 41470125 - 41470537 [+] on Build GRCh38 80
GenBankmRNA BQ771966.1 - 1 Chr. 13 - 41470213 - 41470871 [-] on Build GRCh38 80


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream olfactory receptor family 7 subfamily E member 37 pseudogene OR7E37P OR7E48P, OST193, hg533 -24.4kb Sense Chr. - 41442564 - 41443766 on Build GRCh38
Downstream von Willebrand factor A domain containing 8 VWA8 KIAA0564 96.0kb Anti-sense Chr. - 41566825 - 41961111 on Build GRCh38

View Full Product Details