Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRCH3
Entrez Gene ID
84859
Gene Name
leucine rich repeats and calponin homology domain containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 197790855 - 197889346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005269365.2 XP_005269422.1 20 - 20 9018 95
XM_005269367.2 XP_005269424.1 19 - 19 8946 95
XM_006713791.3 XP_006713854.1 21 - 21 6097 95
XM_006713792.3 XP_006713855.1 20 - 20 6025 95
XM_006713793.2 XP_006713856.1 20 - 20 5989 95
XM_017007352.1 XP_016862841.1 18 - 18 8892 95
GenBank mRNA AK125235.1 - 1 - 1 548 95


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA080686.1 - 1 405 Chr. 3 - 197886602 - 197887161 [+] on Build GRCh38 95
GenBankmRNA DA559512.1 - 1 548 Chr. 3 - 197886457 - 197887271 [+] on Build GRCh38 95
GenBankmRNA BM906317.1 - 168 Chr. - - [-] on Build 95
GenBankmRNA DA380104.1 - 1 487 Chr. 3 - 197886519 - 197887057 [+] on Build GRCh38 95


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream IQ motif containing G IQCG CFAP122, DRC9 Anti-sense Chr. - 197889075 - 197960019 on Build GRCh38
Downstream ribosomal protein L35a RPL35A DBA5, L35A 61.7kb Sense Chr. - 197950181 - 197955851 on Build GRCh38

View Full Product Details