Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ELAVL4
- Entrez Gene ID
-
1996
- Gene Name
- ELAV like neuron-specific RNA binding protein 4
- Gene Aliases
-
HUD, PNEM
- Chromosome Location
-
Chr.1: 50048014 - 50203786 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA161265.1 |
- |
2 |
71 |
|
Chr. 1 - 50109939 - 50193829 [+] on Build GRCh38 |
71 |
GenBankmRNA |
DC352242.1 |
- |
2 |
300 |
|
Chr. 1 - 50108944 - 50145167 [+] on Build GRCh38 |
71 |
GenBankmRNA |
DC397264.1 |
- |
2 |
274 |
|
Chr. 1 - 50119788 - 50177119 [+] on Build GRCh38 |
71 |
GenBankmRNA |
AA214645.1 |
- |
2 |
194 |
|
Chr. 1 - 50109816 - 50145150 [+] on Build GRCh38 |
71 |
GenBankmRNA |
HY024911.1 |
- |
2 |
294 |
|
Chr. 1 - 50119769 - 50145110 [+] on Build GRCh38 |
71 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ATP/GTP binding protein like 4 |
AGBL4 |
CCP6 |
-117.8kb
|
Anti-sense |
Chr. - 48520157 - 50024011 on Build GRCh38 |
Downstream |
DMRT like family A2 |
DMRTA2 |
|
275.7kb
|
Anti-sense |
Chr. - 50417550 - 50423447 on Build GRCh38 |
View Full Product Details