The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027473.1 | 17 - 18 | 3181 | 78 | |
GenBank mRNA | BC047486.1 | - | 17 - 18 | 3183 | 78 |
JQ710743.1 | - | 16 - 17 | 2365 | 78 |