Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INE1
Entrez Gene ID
8552
Gene Name
inactivation escape 1 (non-protein coding)
Gene Aliases
NCRNA00010
Chromosome Location
Chr.X: 47204848 - 47205861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024616.1 1 - 1 553 107
GenBank mRNA BC069671.1 - 1 - 1 476 107
BC069696.1 - 1 - 1 475 107
BC069772.1 - 1 - 1 475 107
BC112149.1 - 1 - 1 365 107
BC112175.1 - 1 - 1 365 107


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AL601447.1 - 1 135 Chr. X - 47205266 - 47206071 [+] on Build GRCh38 107
GenBankmRNA DA392789.1 - 1 194 Chr. X - 47205207 - 47205798 [+] on Build GRCh38 107


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream RNA binding motif protein 10 RBM10 DXS8237E, GPATC9, GPATCH9, S1-1, TARPS, ZRANB5 -18.0kb Sense Chr. - 47145196 - 47186815 on Build GRCh38
Downstream cyclin dependent kinase 16 CDK16 PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1 12.0kb Sense Chr. - 47217881 - 47229997 on Build GRCh38

View Full Product Details