Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C11orf95
Entrez Gene ID
65998
Gene Name
chromosome 11 open reading frame 95
Gene Aliases
-
Chromosome Location
Chr.11: 63759892 - 63768641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144936.1 NP_001138408.1 2 - 3 663 46

View Full Product Details