Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C11orf96
Entrez Gene ID
387763
Gene Name
chromosome 11 open reading frame 96
Gene Aliases
AG2
Chromosome Location
Chr.11: 43942556 - 43943883 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145033.1 NP_001138505.1 1 - 1 551 89
GenBank mRNA BC052560.1 - 1 - 1 517 89

View Full Product Details