Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF035318.1 |
- |
1 |
1346 |
|
Chr. 10 - 11276457 - 11278128 [+] on Build GRCh38 |
92 |
GenBankmRNA |
DB349582.1 |
- |
1 |
|
|
Chr. 10 - 11277551 - 11278110 [-] on Build GRCh38 |
92 |
GenBankmRNA |
BF724324.1 |
- |
1 |
|
|
Chr. 10 - 11277522 - 11278124 [-] on Build GRCh38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
CELF2 antisense RNA 2 |
CELF2-AS2 |
|
-171.0kb
|
Anti-sense |
Chr. - 11071541 - 11105488 on Build GRCh38 |
Downstream |
CELF2 antisense RNA 1 |
CELF2-AS1 |
C10orf31 |
38.7kb
|
Anti-sense |
Chr. - 11316833 - 11344674 on Build GRCh38 |
View Full Product Details