Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW409130.1 |
- |
1 |
70 |
|
Chr. 8 - 11472498 - 11472816 [-] on Build GRCh38 |
66 |
GenBankmRNA |
AF124368.1 |
- |
1 |
950 |
|
Chr. 8 - 11472446 - 11473704 [-] on Build GRCh38 |
66 |
GenBankmRNA |
BU622617.1 |
- |
1 |
|
|
Chr. 8 - 11472440 - 11473140 [+] on Build GRCh38 |
66 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
FAM167A antisense RNA 1 |
FAM167A-AS1 |
C8orf12 |
-33.8kb
|
Anti-sense |
Chr. - 11368402 - 11438657 on Build GRCh38 |
Downstream |
BLK proto-oncogene, Src family tyrosine kinase |
BLK |
MODY11 |
20.3kb
|
Anti-sense |
Chr. - 11493991 - 11564599 on Build GRCh38 |
View Full Product Details