Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AJ276248.1 - 1 55 Chr. 17 - 2460183 - 2460294 [-] on Build GRCh38 93


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream methyltransferase like 16 METTL16 METT10D Sense Chr. - 2416049 - 2511906 on Build GRCh38
Downstream platelet activating factor acetylhydrolase 1b regulatory subunit 1 PAFAH1B1 LIS1, LIS2, MDCR, MDS, NudF, PAFAH 133.3kb Anti-sense Chr. - 2593210 - 2685617 on Build GRCh38

View Full Product Details