Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX357228.2 - 1 Chr. 19 - 1581095 - 1584962 [-] on Build GRCh38 77
GenBankmRNA BU929132.1 - 1 Chr. 19 - 1582395 - 1584931 [-] on Build GRCh38 77
GenBankmRNA DA842237.1 - 1 Chr. 19 - 1584651 - 1584944 [-] on Build GRCh38 77
GenBankmRNA M77818.1 - 1 49 Chr. 19 - 1584735 - 1584925 [-] on Build GRCh38 75
GenBankmRNA BX378302.2 - 1 Chr. 19 - 1578075 - 1584923 [-] on Build GRCh38 77


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream mex-3 RNA binding family member D MEX3D MEX-3D, MEX3, OK/SW-cl.4, RKHD1, RNF193, TINO -16.7kb Sense Chr. - 1554669 - 1568058 on Build GRCh38
Downstream ubiquinol-cytochrome c reductase, complex III subunit XI UQCR11 0710008D09Rik, QCR10, UQCR 12.2kb Sense Chr. - 1597155 - 1605484 on Build GRCh38

View Full Product Details