Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CB963611.1 - 1 Chr. 6 - 143152583 - 143153276 [+] on Build GRCh38 69
GenBankmRNA AF085955.1 - 1 147 Chr. 6 - 143152224 - 143152869 [-] on Build GRCh38 69
GenBankmRNA H69158.1 - 1 147 Chr. 6 - 143152495 - 143152869 [-] on Build GRCh38 69


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 1277 LINC01277 -114.6kb Sense Chr. - 142966421 - 143037582 on Build GRCh38
Downstream adenosine deaminase, tRNA specific 2 ADAT2 DEADC1, TAD2, dJ20N2, dJ20N2.1 270.0kb Sense Chr. - 143422832 - 143450750 on Build GRCh38

View Full Product Details