Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM208B
Entrez Gene ID
54906
Gene Name
family with sequence similarity 208 member B
Gene Aliases
C10orf18, bA318E3.2
Chromosome Location
Chr.10: 5684759 - 5763740 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005252474.3 XP_005252531.1 1 - 1 5240 89
XM_011519515.2 XP_011517817.1 1 - 1 5240 89
XM_011519516.2 XP_011517818.1 1 - 1 5240 89
XM_011519519.2 XP_011517821.1 1 - 1 5240 89
XM_011519520.2 XP_011517822.1 1 - 1 5240 89
XM_017016361.1 XP_016871850.1 1 - 1 5240 89
GenBank mRNA AL049233.1 - 1 - 1 3348 89


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI784109.1 - 1 Chr. 10 - 5698105 - 5698443 [-] on Build GRCh38 89
GenBankmRNA AI280474.1 - 1 Chr. 10 - 5698097 - 5698443 [-] on Build GRCh38 89
GenBankmRNA AA758182.1 - 1 Chr. 10 - 5698009 - 5698443 [-] on Build GRCh38 89


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ankyrin repeat and SOCS box containing 13 ASB13 -28.2kb Anti-sense Chr. - 5638857 - 5666595 on Build GRCh38
Downstream GDP dissociation inhibitor 2 GDI2 HEL-S-46e, RABGDIB 66.8kb Anti-sense Chr. - 5765222 - 5813549 on Build GRCh38

View Full Product Details