Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AL080160.1 - 1 2348 Chr. 20 - 63477931 - 63481310 [+] on Build GRCh38 100


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream potassium voltage-gated channel subfamily Q member 2 KCNQ2 BFNC, EBN, EBN1, ENB1, HNSPC, KCNA11, KV7.2 -5.3kb Anti-sense Chr. - 63400208 - 63472677 on Build GRCh38
Downstream eukaryotic translation elongation factor 1 alpha 2 EEF1A2 EEF1AL, EF-1-alpha-2, EF1A, EIEE33, HS1, MRD38, STN, STNL 6.7kb Anti-sense Chr. - 63488012 - 63499315 on Build GRCh38

View Full Product Details