Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA G30708.1 - 1 152 Chr. 7 - 117000114 - 117000361 [-] on Build GRCh38 99
GenBankmRNA AA077041.1 - 1 152 Chr. 7 - 117000114 - 117000361 [-] on Build GRCh38 99


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ST7 overlapping transcript 4 ST7-OT4 NCRNA00042, ST7OT4 -40.3kb Anti-sense Chr. - 116953899 - 116959813 on Build GRCh38
Downstream microRNA 6132 MIR6132 hsa-mir-6132 19.9kb Anti-sense Chr. - 117020211 - 117020319 on Build GRCh38

View Full Product Details