The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011536097.2 | XP_011534399.1 | 2 - 2 | 6271 | 98 |
GenBank mRNA | AJ420536.1 | - | 1 - 1 | 1352 | 98 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Downstream | ubiquitin protein ligase E3D | UBE3D | C6orf157, H10BH, UBE2CBP, YJR141W | 521.6kb | Anti-sense | Chr. - 82880834 - 83065841 on Build GRCh38 |