Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK022061.1 - 1 1051 Chr. 9 - 19228904 - 19230328 [-] on Build GRCh38 103


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream perilipin 2 PLIN2 ADFP, ADRP -101.3kb Sense Chr. - 19108391 - 19127606 on Build GRCh38
Downstream DENN domain containing 4C DENND4C C9orf55, C9orf55B, RAB10GEF, bA513M16.3 437.0 Anti-sense Chr. - 19230765 - 19374268 on Build GRCh38

View Full Product Details