Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW590303.1 - 1 Chr. 7 - 141780343 - 141780813 [-] on Build GRCh38 110
GenBankmRNA AL832513.1 - 1 1454 Chr. 7 - 141779111 - 141780818 [+] on Build GRCh38 110
GenBankmRNA AI652891.1 - 1 Chr. 7 - 141780249 - 141780812 [-] on Build GRCh38 110
GenBankmRNA AW197234.1 - 1 Chr. 7 - 141780356 - 141780812 [-] on Build GRCh38 110


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream taste 2 receptor member 4 TAS2R4 T2R4 Sense Chr. - 141778489 - 141779388 on Build GRCh38
Downstream taste 2 receptor member 5 TAS2R5 T2R5 9.4kb Sense Chr. - 141790217 - 141791366 on Build GRCh38

View Full Product Details