Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR608159.1 |
- |
1 |
741 |
|
Chr. 18 - 54727344 - 54728860 [+] on Build GRCh38 |
98 |
GenBankmRNA |
BX420746.2 |
- |
1 |
740 |
|
Chr. 18 - 54727344 - 54728328 [+] on Build GRCh38 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
dynactin associated protein |
DYNAP |
C18orf26 |
-127.9kb
|
Sense |
Chr. - 54532713 - 54599544 on Build GRCh38 |
Downstream |
coiled-coil domain containing 68 |
CCDC68 |
SE57-1 |
172.6kb
|
Anti-sense |
Chr. - 54901509 - 54959539 on Build GRCh38 |
View Full Product Details