Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA G36943.1 - 1 254 Chr. 15 - 49607641 - 49608051 [-] on Build GRCh38 112
GenBankmRNA N69668.1 - 1 254 Chr. 15 - 49607641 - 49608051 [-] on Build GRCh38 112


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream family with sequence similarity 227 member B FAM227B C15orf33 Sense Chr. - 49326997 - 49620931 on Build GRCh38
Downstream DTW domain containing 1 DTWD1 MDS009 12.9kb Anti-sense Chr. - 49620980 - 49654448 on Build GRCh38

View Full Product Details