Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK093595.1 - 1 957 Chr. 6 - 156928807 - 156931190 [+] on Build GRCh38 69


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 4466 MIR4466 mir-4466 -149.1kb Anti-sense Chr. - 156779678 - 156779731 on Build GRCh38
Downstream transmembrane protein 242 TMEM242 BM033, C6orf35 357.8kb Anti-sense Chr. - 157289022 - 157324259 on Build GRCh38

View Full Product Details