Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- USP33
- Entrez Gene ID
-
23032
- Gene Name
- ubiquitin specific peptidase 33
- Gene Aliases
-
VDU1
- Chromosome Location
-
Chr.1: 77695987 - 77759879 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK094056.1 |
- |
1 - 1 |
344 |
78 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB257060.1 |
- |
1 |
345 |
|
Chr. 1 - 77697360 - 77697937 [-] on Build GRCh38 |
78 |
GenBankmRNA |
DA362717.1 |
- |
2 |
506 |
|
Chr. 1 - 77697547 - 77701536 [-] on Build GRCh38 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger ZZ-type containing 3 |
ZZZ3 |
ATAC1 |
-12.6kb
|
Sense |
Chr. - 77561526 - 77683440 on Build GRCh38 |
Downstream |
mitoguardin 1 |
MIGA1 |
FAM73A |
81.7kb
|
Anti-sense |
Chr. - 77779624 - 77879540 on Build GRCh38 |
View Full Product Details