Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C16orf72
Entrez Gene ID
29035
Gene Name
chromosome 16 open reading frame 72
Gene Aliases
PRO0149
Chromosome Location
Chr.16: 9091329 - 9119698 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK094351.1 - 1 - 1 949 70


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CA866022.1 - 1 Chr. 16 - 9105731 - 9106246 [-] on Build GRCh38 70
GenBankmRNA CD523126.1 - 1 347 Chr. 16 - 9105452 - 9106160 [+] on Build GRCh38 70


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ubiquitin specific peptidase 7 (herpes virus-associated) USP7 HAUSP, TEF1 -141.4kb Anti-sense Chr. - 8892094 - 8963912 on Build GRCh38
Downstream uncharacterized LOC101927009 LOC101927009 295.0kb Sense Chr. - 9355588 - 9408093 on Build GRCh38

View Full Product Details