Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM199
Entrez Gene ID
147007
Gene Name
transmembrane protein 199
Gene Aliases
C17orf32, CDG2P, VMA12, VPH2
Chromosome Location
Chr.17: 28357581 - 28363683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152464.2 NP_689677.1 6 - 6 3035 70
GenBank mRNA AA279827.1 - 1 - 1 70


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA N95760.1 - 1 Chr. 17 - 28363429 - 28363683 [-] on Build GRCh38 70
GenBankmRNA BX360252.2 - 1 750 Chr. 17 - 28362821 - 28363683 [+] on Build GRCh38 70
GenBankmRNA HY271921.1 - 1 Chr. 17 - 28363249 - 28363682 [-] on Build GRCh38 70
GenBankmRNA AA550719.1 - 1 Chr. 17 - 28363305 - 28363682 [-] on Build GRCh38 70
GenBankmRNA R45602.1 - Chr. - - [-] on Build 70
GenBankmRNA BX360251.2 - 1 Chr. 17 - 28362829 - 28363662 [-] on Build GRCh38 70
GenBankmRNA AI829723.1 - 1 Chr. 17 - 28363296 - 28363672 [-] on Build GRCh38 70
GenBankmRNA AW953506.1 - 341 Chr. - - [-] on Build 70
GenBankmRNA DB334083.1 - 1 Chr. 17 - 28363150 - 28363684 [-] on Build GRCh38 70
GenBankmRNA AI265900.1 - 1 Chr. 17 - 28363301 - 28363683 [-] on Build GRCh38 70
GenBankmRNA BX362448.1 - 1 Chr. 17 - 28362718 - 28363638 [-] on Build GRCh38 70
GenBankmRNA CR613018.1 - 1 750 Chr. 17 - 28362821 - 28363683 [+] on Build GRCh38 70
GenBankmRNA BQ024792.1 - 1 Chr. 17 - 28363035 - 28363683 [-] on Build GRCh38 70
GenBankmRNA BQ018054.1 - 1 Chr. 17 - 28362982 - 28363683 [-] on Build GRCh38 70
GenBankmRNA BM990064.1 - 1 498 Chr. 17 - 28363082 - 28363683 [+] on Build GRCh38 70
GenBankmRNA BF109386.1 - 1 Chr. 17 - 28363199 - 28363683 [-] on Build GRCh38 70
GenBankmRNA DA514504.1 - 1 100 Chr. 17 - 28363471 - 28364015 [+] on Build GRCh38 70
GenBankmRNA R41616.1 - Chr. - - [-] on Build 70
GenBankmRNA AI272163.1 - 1 Chr. 17 - 28363422 - 28363665 [-] on Build GRCh38 70
GenBankmRNA H29880.1 - 1 Chr. 17 - 28363293 - 28363664 [-] on Build GRCh38 70
GenBankmRNA H97168.1 - 1 Chr. 17 - 28363284 - 28363683 [-] on Build GRCh38 70


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 4723 MIR4723 mir-4723 -2.1kb Sense Chr. - 28360654 - 28360734 on Build GRCh38
Downstream SEBOX homeobox SEBOX OG-9, OG9, OG9X 585.0 Anti-sense Chr. - 28364268 - 28365151 on Build GRCh38

View Full Product Details