Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK097036.1 - 1 1241 Chr. 16 - 15745605 - 15747573 [-] on Build GRCh38 73


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream nudE neurodevelopment protein 1 NDE1 HOM-TES-87, LIS4, MHAC, NDE, NUDE, NUDE1 -19.3kb Anti-sense Chr. - 15643267 - 15726353 on Build GRCh38
Downstream FGFR1OP N-terminal like FOPNL C16orf63, FOR20, PHSECRG2 118.1kb Sense Chr. - 15865719 - 15888649 on Build GRCh38

View Full Product Details