Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MNX1
- Entrez Gene ID
-
3110
- Gene Name
- motor neuron and pancreas homeobox 1
- Gene Aliases
-
HB9, HLXB9, HOXHB9, SCRA1
- Chromosome Location
-
Chr.7: 157004853 - 157010653 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY927461.1 |
- |
1 - 1 |
1656 |
76 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BP387239.1 |
- |
1 |
296 |
|
Chr. 7 - 157007597 - 157007931 [-] on Build GRCh38 |
76 |
GenBankmRNA |
BQ269140.1 |
- |
1 |
55 |
|
Chr. 7 - 157007112 - 157007691 [-] on Build GRCh38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MNX1 antisense RNA 2 |
MNX1-AS2 |
|
-419.0
|
Anti-sense |
Chr. - 157005863 - 157007120 on Build GRCh38 |
Downstream |
MNX1 antisense RNA 1 (head to head) |
MNX1-AS1 |
CCAT5 |
1.6kb
|
Anti-sense |
Chr. - 157010857 - 157016424 on Build GRCh38 |
View Full Product Details