Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC098160.1 - 1 357 Chr. 14 - 106745221 - 106745627 [-] on Build GRCh38 73


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream immunoglobulin heavy variable 1-69 IGHV1-69 IGHV1-E, IGHV169, IGHV1E -30.1kb Sense Chr. - 106714682 - 106715120 on Build GRCh38
Downstream immunoglobulin heavy variable 3-71 (pseudogene) IGHV3-71 3-71P, IGHV3-G, IGHV371, IGHV3G 29.5kb Sense Chr. - 106775157 - 106775618 on Build GRCh38

View Full Product Details