Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC105375273
- Entrez Gene ID
-
105375273
- Gene Name
- uncharacterized LOC105375273
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 48708404 - 48711484 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC036696.2 |
- |
1 - 1 |
1045 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ATP binding cassette subfamily A member 13 |
ABCA13 |
|
-61.0kb
|
Sense |
Chr. - 48171424 - 48647497 on Build GRCh38 |
Downstream |
CDC14 cell division cycle 14 C-like |
CDC14BL |
|
134.9kb
|
Anti-sense |
Chr. - 48846402 - 48848081 on Build GRCh38 |
View Full Product Details