Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NCOA2
- Entrez Gene ID
-
10499
- Gene Name
- nuclear receptor coactivator 2
- Gene Aliases
-
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75
- Chromosome Location
-
Chr.8: 70109762 - 70405390 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI669018.1 |
- |
|
358 |
|
Chr. - - [-] on Build |
94 |
GenBankmRNA |
AA338057.1 |
- |
1 |
250 |
|
Chr. 8 - 70111174 - 70111481 [-] on Build GRCh38 |
94 |
GenBankmRNA |
AA382803.1 |
- |
1 |
143 |
|
Chr. 8 - 70111142 - 70111374 [-] on Build GRCh38 |
94 |
GenBankmRNA |
BG034089.1 |
- |
1 |
273 |
|
Chr. 8 - 70110694 - 70111504 [-] on Build GRCh38 |
94 |
GenBankmRNA |
AL048856.1 |
- |
1 |
126 |
|
Chr. 8 - 70110852 - 70111344 [-] on Build GRCh38 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
PR domain 14 |
PRDM14 |
PFM11 |
-38.4kb
|
Sense |
Chr. - 70051126 - 70071327 on Build GRCh38 |
Downstream |
uncharacterized LOC101926892 |
LOC101926892 |
|
359.5kb
|
Anti-sense |
Chr. - 70471134 - 70485687 on Build GRCh38 |
View Full Product Details