Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR743503.1 |
- |
1 |
438 |
|
Chr. 4 - 147491740 - 147492431 [+] on Build GRCh38 |
91 |
GenBankmRNA |
BX537573.1 |
- |
1 |
5346 |
|
Chr. 4 - 147486829 - 147492658 [+] on Build GRCh38 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 548g |
MIR548G |
MIRN548G, hsa-mir-548g |
-142.1kb
|
Anti-sense |
Chr. - 147344629 - 147344717 on Build GRCh38 |
Downstream |
transmembrane protein 184C |
TMEM184C |
TMEM34 |
124.7kb
|
Sense |
Chr. - 147617371 - 147637273 on Build GRCh38 |
View Full Product Details