Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK124863.1 - 1 953 Chr. 12 - 24902309 - 24904681 [+] on Build GRCh38 94


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 477 LINC00477 C12orf67, FAM191B -318.1kb Anti-sense Chr. - 24566964 - 24584168 on Build GRCh38
Downstream chromosome 12 open reading frame 77 C12orf77 88.7kb Anti-sense Chr. - 24993424 - 24997673 on Build GRCh38

View Full Product Details