Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LARP4B
- Entrez Gene ID
-
23185
- Gene Name
- La ribonucleoprotein domain family member 4B
- Gene Aliases
-
KIAA0217, LARP5
- Chromosome Location
-
Chr.10: 806914 - 931900 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK126769.1 |
- |
1 - 1 |
2850 |
100 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CN284204.1 |
- |
1 |
230 |
|
Chr. 10 - 985012 - 985704 [-] on Build GRCh38 |
100 |
GenBankmRNA |
DB300622.1 |
- |
1 |
|
|
Chr. 10 - 985226 - 985635 [+] on Build GRCh38 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105376341 |
LOC105376341 |
|
-28.8kb
|
Sense |
Chr. - 951546 - 956381 on Build GRCh38 |
Downstream |
GTP binding protein 4 |
GTPBP4 |
CRFG, NGB, NOG1 |
80.0
|
Anti-sense |
Chr. - 988409 - 1017768 on Build GRCh38 |
View Full Product Details