Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK130660.1 |
- |
1 |
2352 |
|
Chr. 21 - 34863573 - 34866308 [-] on Build GRCh38 |
83 |
GenBankmRNA |
BQ709986.1 |
- |
1 |
|
|
Chr. 21 - 34863705 - 34864517 [-] on Build GRCh38 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1426 |
LINC01426 |
lincRNA-uc002yug.2 |
-78.7kb
|
Anti-sense |
Chr. - 34745825 - 34784886 on Build GRCh38 |
Downstream |
RUNX1 intronic transcript 1 |
RUNX1-IT1 |
C21orf96 |
171.6kb
|
Sense |
Chr. - 35037925 - 35039426 on Build GRCh38 |
View Full Product Details